VWR® Drosophila Vial Plugs, Cellulose Acetate
Catalog # 89168-886
Supplier: VWR International
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Specifications
- Description:Fits 25 x 95 mm Vials
- Cat. no.:89168-886
Specifications
About this item
Bonded dense-weave cellulose acetate plugs provide a better barrier to mites with minimal shedding.
- Cellulose acetate fabrication
- Excellent barrier to mites
- Minimal shedding
Reduce evaporation to keep food fresh longer.
Customers Who Bought This Also Bought