ContactUsLinkComponent
C & S VIALS
Protocol™ Parasitology Vials, Thermo Scientific
# de catalogue 89167-488
Protocol™ Parasitology Vials, Thermo Scientific
# de catalogue 89167-488
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    C&S Vials
  • Cat. no.:
    89167-488
  • Supplier No.:
    23-005-43
Spécifications
A propos de cet article
For the collection, transport, and examination of fecal specimens
Sodium acetate-acetic acid-formalin (SAF) is ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, and cocciadian oocysts and microsporidian spores. Slides fixed with SAF may be stained using trichrome or hematoxylin.
For the collection, transport, and examination of fecal specimens
Sodium acetate-acetic acid-formalin (SAF) is ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, and cocciadian oocysts and microsporidian spores. Slides fixed with SAF may be stained using trichrome or hematoxylin. 10% neutral buffered formalin is an all purpose fixative appropriate for helminth eggs and larvae and for protozoan cysts. The buffering system helps to maintain organism morphology. Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae, and protozoan cysts.
Sodium acetate-acetic acid-formalin (SAF) is ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, and cocciadian oocysts and microsporidian spores. Slides fixed with SAF may be stained using trichrome or hematoxylin. 10% neutral buffered formalin is an all purpose fixative appropriate for helminth eggs and larvae and for protozoan cysts. The buffering system helps to maintain organism morphology. Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae, and protozoan cysts.
C&S media and Cary Blair media facilitate the collection and transport of enteric bacterial pathogens, while maintaining the bacterial population for optimum recovery for up to 96 hours. C&S medium: Agar, sodium thioglycolate, Na2HPO4, calcium chloride, phenol red. Cary Blair medium: Agar, DI water, KH2PO4, NA2HPO4, phenol red, sodium chloride, sodium thioglycolate.
Modified (Cu) PVA fixative acts as a preservative and fixative ideally suited for protozoan trophozoites. Slides fixed with Modified PVA may be stained with trichrome or hematoxylin.
Zn-PVA is used as a preservative and fixative for protozoan trophozoites. Proper use assures the parasitologist that diagnostic stages of intestinal parasites, if present, will be preserved. Slides fixed Zn-PVA may be stained using trichrome or hematoxylin.
A full line of mercury-free fixatives and preservatives are available in 15mL single vials, or double- and triple-vial kits to meet individual laboratory needs
Ordering information: Single-vial kits contain 20 vials. Double- and triple-vial kits contain ten vials of each type.
Customers Who Bought This Also Bought