Order Entry
Canada
ContactUsLinkComponent
Round German Glass Cover Slips, Chemglass
Catalogue # 89167-106
Fournisseur:  Chemglass
Small Business Enterprise
undefined
Round German Glass Cover Slips, Chemglass
Catalogue # 89167-106
Fournisseur:  Chemglass
Supplier Number:  CLS-1760-012
Small Business Enterprise

Spécifications

  • Ø:
    12 mm
  • Description:
    Cover slips, round
  • Packed:
    1 oz. package, approx. 650 slides/oz.
  • Cat. no.:
    89167-106
  • Supplier No.:
    CLS-1760-012

Spécifications

À propos de cet article

Cover slips are manufactured from the highest quality hydrolytic Class 1 German borosilicate glass

All cover slips are #1 thickness. They have excellent resistance to chemicals and are ideal for neural tissue culture and hard-to-attach cells. These cover slips are exceptional for florescence and confocal microscopy. Sold in 1 oz. packages.

Frequently Bought Together