ContactUsLinkComponent
Vial Only Sample, 8Ml, Amber, Gpi 15-425, 17 X 60Mm
# de catalogue 89166-686
Fournisseur:  Chemglass
Vial Only Sample, 8Ml, Amber, Gpi 15-425, 17 X 60Mm
Vial Only Sample, 8Ml, Amber, Gpi 15-425, 17 X 60Mm
# de catalogue 89166-686
Fournisseur:  Chemglass
Customers Who Bought This Also Bought