Order Entry
ContactUsLinkComponent
Vials
Sample Vials, Clear, Chemglass
# de catalogue 89166-660
Fournisseur:  Chemglass
Small Business Enterprise
Vials
Sample Vials, Clear, Chemglass
# de catalogue 89166-660
Fournisseur:  Chemglass
Small Business Enterprise

Spécifications

  • Cap included:
    No
  • Cap Size:
    13-425
  • Capacity:
    4.00 ml
  • Color:
    Clear
  • Cat. no.:
    89166-660
  • Diameter ×H:
    15×45 mm
  • Supplier No.:
    CG-4902-02

Spécifications

A propos de cet article

Clear sample vials are made from Type 1 borosilicate glass.

Packaging: Sample vials packaged in convenient LAB-PACS.
Customers Who Bought This Also Bought