ContactUsLinkComponent
VWR Disposable Storage Bottles
VWR® Round Media Bottles
# de catalogue 89166-094
Fournisseur:  VWR International
VWR Disposable Storage Bottles
VWR® Round Media Bottles
# de catalogue 89166-094
Fournisseur:  VWR International
Spécifications
  • Capacity:
    500 ml (16.9 oz.)
  • Sterility:
    Sterile
Spécifications
A propos de cet article
Disposable round media bottles for storing sterile solutions such as tissue culture media, serum, and buffers.
  • Polycarbonate or polystyrene construction
  • Sloped bottle design
  • Easy to grip
  • Easily visible graduations
Bottles feature a sloped design that improves stability on the work surface.
Polycarbonate bottles can withstand up to three autoclave cycles and offer good alcohol resistance. They are stable up to 135 °C (275 °F). Polystyrene bottles are ideal for general-purpose cell culture aqueous solutions, and are stable up to 80 °C (176 °F). PETG bottles are ideal for most general laboratory solutions.
The narrow bottle shoulder and molded finger grips enable the bottle to be gripped securely by gloved hands. Polypropylene caps feature a and convenient gripping surface for easy handling and transporting. Caps allow for easy uncapping, tightening, and bottle stacking. Tops of caps feature a flat writing surface.
Packaging: Bottles are double wrapped in packaging.
Customers Who Bought This Also Bought