ContactUsLinkComponent
Biomol Green* Reagent 250 Ml
BIOMOL® Green, Enzo Life Sciences
# de catalogue 89156-424
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Biomol Green* Reagent 250 Ml
BIOMOL® Green, Enzo Life Sciences
# de catalogue 89156-424
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    250 mL
  • Cat. no.:
    89156-424
  • Supplier No.:
    AK111-0250
Spécifications
A propos de cet article
BIOMOL Green™ provides a simple and convenient method for colorimetric phosphate quantitation (abs. 600-680 nm). Unlike other molybdate/malachite green-based assays, BIOMOL Green™ doesn’t require multiple solutions or reagents prepared fresh on the day of the assay. BIOMOL Green™ is extremely stable (>6 mos. at 4°C) and is simply mixed, at room temperature, with any enzymatic reaction that has released free phosphate. Reported applications include assays for phospholipid phosphatases, tyrosyl-tRNA synthetase (coupled with pyrophosphatase) and a viral RNA triphosphatase.BIOMOL Green™ has perhaps been most widely applied to protein phosphatase assays. Typically, a protein phosphatase (e.g. PTP1B or LAR) is simply incubated with an appropriate phosphopeptide (e.g. IR5,9,10). Addition of BIOMOL Green™ stops the reaction and begins color development, which is read 20-30 min. later.BIOMOL Green™ may be used in cuvette or microplate-based assays and is ideal for high-throughput applications. Each kit comes with a standardized phosphate solution for assay calibration.
Ordering information: Contents: Biomol Green™ Reagent (Prod. No. BML-AK111) (1l or 250ml; liquid in screw cap plastic bottle) Storage: room temperature. Long-term at 4°C; can be stored frozen without deleterious effects.
Phosphate Standard (Prod. No. BML-KI102) (1ml; 800µm phosphate in distilled water) Storage: room temperature. Long-term at 4°C; can be stored frozen without deleterious effects.
Customers Who Bought This Also Bought