VWR® Four-Channel Economy Timer
Catalog # 89140-552
Supplier: VWR International
Specifications
- Description:Economy 4-channel timer
- Cat. no.:89140-552
Specifications
About this item
Time-of-day clock.
- Loud alarm
- Memory function remembers the last time set
- Fold-out stand and magnet included
Counts up or down simultaneously for 99 hours, 59 minutes, 59 seconds.
Customers Who Bought This Also Bought