ContactUsLinkComponent
VWR Economy Digital Thermometer
VWR® Economy Digital Thermometer
# de catalogue 89140-510
Fournisseur:  VWR International
VWR Economy Digital Thermometer
VWR® Economy Digital Thermometer
# de catalogue 89140-510
Fournisseur:  VWR International
Spécifications
  • Resolution:
    0.1
  • Description:
    VWR® Digital Thermometer with Probe
  • Probe:
    Sensor with 39" immmersible cable
  • Range:
    –10...50 °C (14...122 °F), ambient; –50...70 °C (–58...158 °F), probe
Spécifications
A propos de cet article
Economy digital thermometer with probe and used in ambient temperature.
  • High or low alarm throughput
  • With minimum and maximum memory
  • Available in °C and °F
Customers Who Bought This Also Bought