ContactUsLinkComponent
VWR*Slide Box Tall
VWR® Tall Microscope Slide Boxes for 100 Slides
# de catalogue 89140-280
Fournisseur:  VWR International
VWR*Slide Box Tall
VWR® Tall Microscope Slide Boxes for 100 Slides
# de catalogue 89140-280
Fournisseur:  VWR International
Spécifications
  • Dimensions:
    208W x 175H x 60D mm (8⅛ x 6⅞ x 2⅜")
  • Color:
    Blue
  • Description:
    VWR® Tall Microscope Slide Boxes, 100-Place
  • Cat. no.:
    89140-280
Spécifications
A propos de cet article
Tall slide boxes are designed for storage and transport of non-standard microscope slides (3x2").
  • Grooved slots
  • Soft cork lining
  • Sturdy ABS construction for rugged use
Grooved slots hold individual slides in place. The box lid provides the needed clearance for the taller height of non-standard slides. Soft cork lining insures that your slides will not slip. Sturdy ABS construction for rugged use.
Customers Who Bought This Also Bought