Spécifications
- Description:Flashing LED alert big-digit dual channel timer
- Cat. no.:89140-196
- W×H×D:3×3¹/₂×⁵/₈"
Spécifications
A propos de cet article
Allows simultaneous programming of two different activities from 99 hours, 59 minutes, 59 seconds to 1 second.
- Bright LED digits
- Memory function for repetitive timing
- Unique visual indicator during countdown
Resolution is 1 second; accuracy is 0.01%
For repetitive timing, the microcomputer chip permits setting 2 countdown times into memory. The memory recalls the last countdown time set. Large ⁵/₈-inch high LCD digits may be read from 18 feet.
Unique visual indicator for each channel when counting down, green light flashes every second once the countdown time reaches 10 seconds. Red light flashes when zero has been reached and the alarm is sounding. Alarm has three positions which allow setting to high decibel for noisy areas, low decibel for quiet environments, or off the muted setting for silent, visual flashing LED-only operation.
Each channel has a distinct electronic alarm and visual indicator when zero is reached. Audible alarms turns off automatically after one minute or may be silenced automatically.