ContactUsLinkComponent
GelGreen* Nucleic Acid Stain
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
# de catalogue 89139-150
Fournisseur:  Biotium
   Safer Human and Environmental Health
Numéro CAS:  
GelGreen* Nucleic Acid Stain
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
# de catalogue 89139-150
Fournisseur:  Biotium
   Safer Human and Environmental Health
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    GelGreen™ 10000X in water (trial)
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Size:
    100 μl (trial size)
  • Environmental certification:
    Classified as non-hazardous to aquatic life under CCR title 22 regulations
  • Cat. no.:
    89139-150
  • Supplier No.:
    41005-T
Spécifications
A propos de cet article
GelRed™ and GelGreen™ are a new generation of fluorescent nucleic acid gel stains designed to replace the highly toxic ethidium bromide (EtBr)
GelRed™ and GelGreen™ use a novel yet very simple concept to reduce genotoxicity by preventing the dyes from entering living cells. A DNA-binding dye can be made nonmutagenic or substantially less mutagenic by denying contact with genomic DNA in living cells. Thus, GelRed™ and GelGreen™ are engineered such that the chemical structures of the dyes are incapable of crossing cell membranes. The Ames test confirmed that GelRed™ and GelGreen™ are nonmutagenic at concentrations well above their working concentrations used for gel staining. Furthermore, environmental safety tests showed that GelRed™ and GelGreen™ are nonhazardous and nontoxic to aquatic life. As a result, GelRed™ and GelGreen™ can be disposed of down the drain or in regular trash.
GelRed™ and GelGreen™ are highly sensitive either as precast gel stains or post gel stains. Designed primarily for use with a 312/302 nm UV transilluminator, GelRed is much more sensitive than EtBr. GelRed™ can also be used as a highly sensitive precast gel stain. GelGreen™ is developed to meet the needs of researchers who use a 488 nm laser-based gel scanner or a Dark Reader that uses a visible blue light for excitation. The dyes are stable in water at room temperature for long-term storage, and can be microwaved for making precast gels. Both dyes are also very photostable allowing use under normal room lighting.
GelRed and GelGreen are superior to EtBr and other EtBr alternatives by having a combination of low toxicity, high sensitivity and exceptional stability.
Customers Who Bought This Also Bought