ContactUsLinkComponent
The AccuBlue Broad Range dsDNA Quantitation Solution
AccuBlue® and AccuClear® dsDNA Quantitation Kits, Biotium
# de catalogue 89139-018
Fournisseur:  Biotium
Numéro CAS:  
The AccuBlue Broad Range dsDNA Quantitation Solution
AccuBlue® and AccuClear® dsDNA Quantitation Kits, Biotium
# de catalogue 89139-018
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    AccuBlue® Broad Range dsDNA quantitation solution, trial size
  • Size:
    200 assays
  • Cat. no.:
    89139-018
  • Supplier No.:
    31009-T
Spécifications
A propos de cet article
AccuBlue™ and AccuClear™ dsDNA quantitation assays provide sensitive, accurate and easy-to-use dsDNA quantitation assays for fluorescent microplate readers in 96-well or 384-well formats.
  • Accurately measure double-stranded DNA from a wide range of samples, from low picogram to microgram
  • Quantify many samples and replicates at once for better accuracy and high-throughput analysis in 96-well or 384-well formats
  • Compatible with a range of instruments including fluorescence microplate readers, spectrofluorometers including Qubit®, QuantiFluor-P®, or Nanodrop®, and other hand-held fluorometers
  • Available in a wide selection of kit choices for a wide range of samples
AccuBlue® and AccuClear® dsDNA quantitation assays allow precise quantitation of purified dsDNA samples across a wide range of concentrations and a variety of fluorescence detection instruments. Unlike absorbance-based nucleic acid quantitation, fluorescent DNA binding dyes are highly sensitive and selective for double-stranded DNA and provide a more accurate DNA concentration in the presence of contaminating RNA and other common contaminants including free nucleotides, protein, detergents and salts. Some of our kits offer unrivaled sensitivity or linear ranges. To choose the DNA quantitation assay that is right for you, see the linear detection ranges and features in the table.
AccuBlue® Broad Range Assay provides a wide dynamic range of 2 to 2000 ng per well of a 96 well plate (0.2 to 200 ng/µl of sample). The standard curve can be extended to 4000 ng/well with slight compromise in linearity. Excitation/emission wavelengths: 350/460 nm.
AccuBlue® High-sensitivity Assay has a linear range of 0.2 to 100 ng dsDNA per well of a 96 well plate (0.02 to 10 ng/µl of sample). Excitation/emission wavelengths: 500/530 nm. The dye is non-toxic and non-mutagentic for safe handling and easy disposal.
AccuClear® Ultra High Sensitivity Assay is extremely versatile, with a very broad dynamic range 30 pg to 250 ng dsDNA per well of a 96-well plate, allowing quantitation of nearly any sample. Excitation/emission wavelengths: 468/507 nm.
AccuBlue® NextGen Assay is the most sensitive DNA quantitation assay on the market, allowing accurate detection of 1 pg of DNA (depends on the sensitivity of the instrument), and rivaling PicoGreen® in sensitivity and accuracy. Excitation/emission wavelengths: 468/507 nm. Specially designed for scientists who need to quantify extremely dilute or precious DNA, ideal for use in preparing samples for applications such as Next Generation Sequencing (NGS) and digital PCR.
Ordering information: AccuBlue™ and AccuClear™ kits include quantitation solution and calf thymus dsDNA standards. Stand-alone AccuBlue™ and AccuClear™ quantitation solutions are available for users who prefer to use their own DNA standards. Calf thymus dsDNA standard sets are also available as stand-alone products.
Customers Who Bought This Also Bought