ContactUsLinkComponent
Pig Blue Heavweight Mat Rolls 15Inx150Ft
Pig Blue® Absorbent Mat Roll, New Pig
# de catalogue 89132-778
Fournisseur:  New Pig
Pig Blue Heavweight Mat Rolls 15Inx150Ft
Pig Blue® Absorbent Mat Roll, New Pig
# de catalogue 89132-778
Fournisseur:  New Pig
Spécifications
  • Description:
    Heavyweight
  • Cat. no.:
    89132-778
  • Dimensions:
    38.1 cm x 46 m (15" x 150')
  • Supplier No.:
    BLU103
Spécifications
A propos de cet article
Consistent construction produces a mat that's free of light spots to absorb liquid evenly for better wicking and retention
Contains 75% recycled fibers; ideal in situations where green products are desired or required. Fire retardant for added safety; flame-tested in accordance with ASTM Standard E84-08. Absorbs oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids. Rolls are ideal for covering large areas and soaking up leaks and overspray. Easy-tear perforations let you take only what you need, so you use less mat and save money.
Heavyweight Pig Blue construction is super absorbent for your most demanding tasks and large-volume cleanups.
FiberFusion Technology fuses superabsorbent natural fibers with strong synthetic fibers for one high-performance absorbent. Stays together, even when picking up or shifting a fully saturated mat.
Customers Who Bought This Also Bought