ContactUsLinkComponent
Humidity Indicator Cards
Humidity Indicator Card, Desco
# de catalogue 89131-360
Fournisseur:  Desco
Product available on GSA Advantage®
Humidity Indicator Cards
Humidity Indicator Card, Desco
# de catalogue 89131-360
Fournisseur:  Desco
Product available on GSA Advantage®
Spécifications
  • Description:
    Humidity Indicator Card
  • Cat. no.:
    89131-360
  • Supplier No.:
    13870
Spécifications
A propos de cet article
Reversible Humidity Indicator Cards for vapor protection measure the level of humidity that the contents in package are exposed.
  • For use with Statshield® ESD Shielding Moisture Barrier Bags
  • Humidity indicator spots are reversible
  • 100 cards per can
  • Made in the United States of America
Cards measure the level of humidity that the contents of the packaging have been exposed to. They can be used with Statshield® ESD Shielding Moisture Barrier Bags to provide protection for moisture sensitive items such as surface mount devices. Reversible design allows the card to be reused, providing better value.
Certifications: Meets MIL-I-8835A And JEDEC J-STD-033D (Type 1) Table 3-1 Dry Packing requirements. Lead-Free, RoHS compliant.
Customers Who Bought This Also Bought