ContactUsLinkComponent
VWR Hand Care, touch free, MicroDispenser
VWR® MicroDispenser™ Touch-Free Soap Dispenser
# de catalogue 89128-812
Fournisseur:  VWR International
Small Business Enterprise
VWR Hand Care, touch free, MicroDispenser
VWR® MicroDispenser™ Touch-Free Soap Dispenser
# de catalogue 89128-812
Fournisseur:  VWR International
Small Business Enterprise
Spécifications
  • Color:
    White
  • Description:
    Touch-free dispenser
  • Cat. no.:
    89128-812
Spécifications
A propos de cet article
Touch-free soap dispensing unit effectively reduces cross-contamination and waste in the cleanroom and gowning areas. The dispenser with drip tray is easily mounted to a wall or floor stand.
  • Reduces cross-contamination and waste in the cleanroom and gowning areas
  • Activated by an electronic eye
  • Delivers a consistent, premeasured dose of soap, lotion, or sanitizer into the palm
  • Gravity fed with no tubes or pumps
  • No-drip unit may be wall-mounted
  • Complies with OSHA, CDC, and ADA requirements
Easy delivery system allows dispenser to be use for different soaps or lotions by changing spout.
Activated by an electronic eye, the dispenser delivers a consistent, premeasured dose of soap, lotion, or sanitizer into the palm of the hand cleanly, safely, and economically.
Gravity fed with no tubes or pumps. A capsule is screwed onto the soap bottle which is then placed upside down on top of the dispenser. Easy delivery system makes the unit available for different soaps and sanitizers by changing the spout from green to blue. No-drip unit may be wall-mounted.
Certifications: Complies with OSHA, CDC, and ADA requirements.
Ordering information: Operates on four size AA batteries (included). Can be used with Micronova 500 ml (16.9 oz.) pump bottles.
Customers Who Bought This Also Bought