Order Entry
ContactUsLinkComponent
CS40 CELLVIEW CLTRDISH 1W ST TC
CELLview™ Cell Culture Dishes, Glass Bottom, Sterile, Greiner bio-one
Catalog # 89125-438
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
CS40 CELLVIEW CLTRDISH 1W ST TC
CELLview™ Cell Culture Dishes, Glass Bottom, Sterile, Greiner bio-one
Catalog # 89125-438
Supplier:  Greiner Bio-One
Product available on GSA Advantage®

Specifications

  • Color:
    Natural
  • Cat. no.:
    89125-438
  • Working Volume:
    5 ml
  • Growth Area:
    8.7 cm2 Per Compartment
  • No. of Compartments:
    1
  • Surface Treatment:
    Tissue culture (TC)
  • Supplier No.:
    627860
  • Total Volume:
    10 ml

Specifications

About this item

The CELLview™ cell culture dish is made from high-grade polystyrene combinded with an integrated glass bottom.

  • Sterilized by irradiation
  • Free of RNase, DNase, and human DNA
  • Noncytotoxic
  • Nonpyrogenic

The innovative design guarantees a single-plane, flat bottom with a consistent working distance and maximal planarity

CELLview™ combines the conveniance of a standard size 35mm disposable plastic cell culture dish with the optical quality of glass, providing superior high-resolution microscopic images of in-vitro cultivated cultures.

Glass bottom features high transparent achromatic borosilicate glass; hydrolytic class 1 (DIN ISO 719) Glass thickness 175 um +/- 15um. Manufactured according to ISO 8255-1; 1986

The subdivided version of the CELLviewCell Culture Dish enables simultaneous multiplex analyzes of different cell lines, varioius stimulations, or diverse transfections.

Ordering information: Sterile and user friendly packaging. Glass lid.
Customers Who Bought This Also Bought