Axygen® Disposable Reagent Reservoirs, Corning
# de catalogue 89108-014
Fournisseur: Corning
Spécifications
- Capacity:100 ml
- Sterility:Sterile
- Description:Disposable Reagent Reservoir
- Environmentally Preferable:Low Manufacturing Impact
- Material:Plastic
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:89108-014
- Packaging:200/case
- Supplier No.:RES-V-100-S
Spécifications
A propos de cet article
These disposable reagent reservoirs are sterile and certified nonpyrogenic
They feature a sharp-V bottom that allows efficient extraction of the reagent. They also feature four pour-off corners and a stacking lug for easy separation
Customers Who Bought This Also Bought