Spécifications
- Capacity:—
- Sterility:Non sterile
- Description:Lid
- Packaging:Bulk
Spécifications
A propos de cet article
Sturdy, shatter-resistant containers are manufactured from transparent polypropylene to allow constant sample viewing.
- Sturdy, shatter-resistant design
- Transparent siding
- Polypropylene construction
- Available in nonsterile or sterile formats
Sturdy, shatter-resistant containers are manufactured from transparent polypropylene to allow constant sample viewing.
Containers feature molded graduations in 10 mL and 0.5 oz. increments. Polyethylene snap-on lids are sold separately, except where noted. Lids create a secure seal, and offer space for date, name, doctor, and room number molded into the surface. Components are available in nonsterile or sterile formats.