VWR® Labels 'Radioactive Warning'
# de catalogue 89098-012
Fournisseur: VWR International
Spécifications
- Description:Warning label with notation spaces
- Cat. no.:89098-012
- Dimensions:–
Spécifications
A propos de cet article
Labels are designed to aid in identifying equipment, containers, rooms, and other items that contain radiation or contaminated agents.
The yellow labels are imprinted with "CAUTION RADIOACTIVE MATERIAL" in distinctive magenta ink to attract immediate attention. Label 89098-012 also includes spaces for noting Isotope, Amount, and Date.
- Prominent radioactive warning imprint
- Space for additional notes
- Distinctive coloring to attract immediate attention
Labels are designed to aid in identifying equipment, containers, rooms, and other items that contain radiation or contaminated agents.
Labels meet various state and federal regulations.
The yellow labels are imprinted with 'CAUTION RADIOACTIVE MATERIAL' in distinctive magenta ink to attract immediate attention. Label 89098-012 also includes spaces for noting Isotope, Amount, and Date.
Ordering information: Supplied as a single 12.7 m (41.7') roll on a 2.5 cm (1") Ø core.
Customers Who Bought This Also Bought