VWR® Hi-Lo Temperature Tape
1.9 cm (¾")
Catalog # 89097-994
Supplier: VWR International
Jump to:
Product Details & Documents
Temperature tape is designed for cryogenic labeling procedures, as well as for use in ovens, sterilizers, and incubators.
- Heat- and cold-resistant tape
- Self-adhesive
- Textured surface for writing
The tape can withstand temperatures up to 287 °C (549 °F) and as low as -146 °C (-232 °F). White tape features a textured surface that can be stamped or written on with a carbon pencil. Length: 7.62 m (25 ft.).
Documents
Certificates
Recherche par numéro de lotSpécifications
- Largeur:1.9 cm (¾")