VWR® Hi-Lo Temperature Tape | Avantor
VWR® Hi-Lo Temperature Tape
1.9 cm (¾")
Catalog # 89097-994
Supplier:   VWR International
VWR® Hi-Lo Temperature Tape
Catalog # 89097-994
Supplier:  VWR International
VWR® Hi-Lo Temperature Tape
VWR® Hi-Lo Temperature Tape
1.9 cm (¾")
Price
63,98 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1
Product Details & Documents
Temperature tape is designed for cryogenic labeling procedures, as well as for use in ovens, sterilizers, and incubators.
  • Heat- and cold-resistant tape
  • Self-adhesive
  • Textured surface for writing
The tape can withstand temperatures up to 287 °C (549 °F) and as low as -146 °C (-232 °F). White tape features a textured surface that can be stamped or written on with a carbon pencil. Length: 7.62 m (25 ft.).
Documents
Spécifications
  • Largeur:
    1.9 cm (¾")
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.