VWR® Red Border Tape Labels
Catalogue # 89097-978
Fournisseur: VWR International
Spécifications
- Description:Border tape labels
- Dimensions:25.4×19.1 mm (1×0.75")
- Cat. no.:89097-978
Spécifications
À propos de cet article
Designed to aid in the identification of containers, shelves, and other standard laboratory or workplace items.
- Self-adhesive tape design
- Distinctive coloring for quick identification
Labels are printed on a white. These labels can be written on with pen, pencil, or permanent marker. They can be easily removed and will not leave a sticky residue.
Ordering information: Supplied on individual rolls, each containing 12.7 m (41.7') of tape.
Frequently Bought Together