VWR® Write-On Labels with Protective Cover
Catalog # 89097-892
Supplier: VWR International
Specifications
- Description:Write-on hazard warning labels
- Dimensions:24×50.8 mm (0.94×2")
- Cat. no.:89097-892
Specifications
About this item
Radiant orange write-on labels with a protective cover prevent the loss of valuable information.
- No dispenser required
- Easy and simple application
Designed for labeling reagents, samples, or noting calibration data.
Labels protect written information from tampering, moisture, smearing, heat and cold, chemicals, and solvents.
Simply write information on label, peel backing from transparent film, and press film over the label. Apply the label to any clean, dry surface.
Packaging: 500 labels per roll.
Customers Who Bought This Also Bought