VWR® Biohazard Labels
# de catalogue 89097-876
Fournisseur: VWR International
Spécifications
- Description:Biohazard labels
- Cat. no.:89097-876
- Dimensions:127×127 mm (5×5")
Spécifications
A propos de cet article
Labels aid in general workplace safety and efficiency by identifying contaminated equipment containers, rooms, or materials.
- Quick and easy identification
- Bold symbols declare biohazard warning from a distance
A biohazard symbol along with the term are printed in bold black atop a fluorescent red background.
Certifications: Labels are manufactured in accordance with various federal and state regulations, including OSHA safety standards.
Ordering information: Labels are available on rolls of 250 or 500 labels. One roll per case.
Customers Who Bought This Also Bought