KIMBLE® KimCote® GL 45 Media Bottles, DWK Life Sciences
# de catalogue 89097-376
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Spécifications
- Capacity:2 L (67.6 oz.)
- Graduation Interval:100 ml
- Graduation range:400 – 1800 mL
- Height:260 mm
- Cat. no.:89097-376
- Outer Diameter:136 mm
- Supplier No.:KC14395-2000
Spécifications
A propos de cet article
Ideal for general laboratory use including mixing, storing or transporting culture media, chemicals, or solvents.
- 30 mm Int.Ø. opening
- Blue polypropylene linerless GL 45 screw thread cap
- Enhanced graduations and marking spot made with chemically resistant white enamel paint
Autoclavable KimCote® safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Blue caps and clear pour rings included.
Customers Who Bought This Also Bought