Order Entry
ContactUsLinkComponent
Stir Shaft
Heavy Duty Polished Glass Stirrer Shaft, 28 mm, Ace Glass Incorporated
# de catalogue 89097-174
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Stir Shaft
Heavy Duty Polished Glass Stirrer Shaft, 28 mm, Ace Glass Incorporated
# de catalogue 89097-174
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Spécifications

  • Fits Flask:
    50 low stance flasks
  • Shaft Length:
    40"
  • Supplier No.:
    8080-12

Spécifications

A propos de cet article

Heavy duty, 28 mm O.D

For Agitators and Turbines Reference the Following Numbers: 89062-152, 89062-174, 89062-176, 71001-576, 71001-578.

glass stirring shaft for use with ACE reactors. Shaft has pre-drilled holes at top and bottom for positive attachment to motor coupling and agitator, respectively. Recommended for use with ACE PTFE Trubore® and mechanical seal bearings. Use with agitators for 28 mm rods. Fits Ace 89054-598 swivel connector and 89054-602 chuck.

Customers Who Bought This Also Bought