Nalgene® Polypropylene Economy Bottles, Wide Mouth, Thermo Scientific
Catalog # 89096-766
Supplier: Thermo Fisher Scientific
Specifications
- Cap size:38 mm
- Ø:50.5 mm
- Capacity:125 ml (4.2 oz.)
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Cat. no.:89096-766
- Supplier No.:2187-0004
- Height with Cap:100.8 mm
- Height without Cap:98.3 mm
Specifications
About this item
Nalgene® Economy Bottles are lightweight, more flexible versions of the original Nalgene Lab Quality Bottles for less rigorous lab use.
- Bottles are autoclavable, guaranteed leakproof
- made in our ISO-certified USA manufacturing facility
These Nalgene® Economy Bottles feature excellent impact and chemical resistance and clarity, include a polypropylene closure, and are made from natural, translucent polypropylene copolymer.
Priced below low-quality plastic or glass bottle brands, Nalgene® Economy Bottles are the economical, quality choice for lighter-duty lab use.
These bottles are suitable for light-duty, general purpose laboratory applications such as containing samples and daily working solutions like buffers and bench reagents.
Ordering information: Thermo Scientific Nalgene® High-Density Polyethylene Environmental Sample Bottles are also available; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought