Nalgene® Polypropylene Economy Bottles, Narrow Mouth, Thermo Scientific
# de catalogue 89096-756
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:24-415 mm
- Capacity:250 ml (8.5 oz.)
- Ø:61 mm
- Cat. no.:89096-756
- Supplier No.:2087-0008
- Height with Cap:132.8 mm
- Height without Cap:130.6 mm
Spécifications
A propos de cet article
Nalgene® Economy Bottles are lightweight, more flexible versions of the original Nalgene Lab Quality Bottles for less rigorous lab use.
- Bottles are the economical, quality choice for lighter-duty lab use
- Priced below low-quality plastic or glass bottle brands
These Nalgene® Economy Bottles feature excellent impact and chemical resistance and clarity, include a polypropylene closure, and are made from natural, translucent polypropylene copolymer. Like most Nalgene bottles, the Economy Bottles are autoclavable, guaranteed leakproof and are made in our ISO-certified USA manufacturing facility.
These bottles are suitable for light-duty, general purpose laboratory applications such as containing samples and daily working solutions like buffers and bench reagents.
Ordering information: Thermo Scientific Nalgene High-Density Polyethylene Environmental Sample Bottles are also available; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought