Order Entry
ContactUsLinkComponent
VWR* Traceable* 5ml Vaccine Thermometer
VWR® Traceable® Refrigerator/Freezer Digital Thermometer
Catalog # 89094-778
Supplier:  VWR International
VWR* Traceable* 5ml Vaccine Thermometer
VWR® Traceable® Refrigerator/Freezer Digital Thermometer
Catalog # 89094-778
Supplier:  VWR International

Specifications

  • Description:
    5 ml Vaccine Thermometer
  • Temperature resolution:
    1°C
  • Temperature accuracy:
    ±1°C
  • Cat. no.:
    89094-778
  • Probe:
    5 ml vaccine bottle probe (glycol)
  • Range:
    –50…+70 °C (–58…+158 °F)

Specifications

About this item

Accurately monitor temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with this enclosed temperature-buffered sensor.

  • Probe is sealed in glycol solution
  • Min/max memory
  • High and low settable alarms

Bottle insulates sensor from rapid temperature changes when refrigerator door is opened.

Triple display simultaneously shows high, low, and current temperatures. Min/max monitors high/low readings overnight, on weekends, or for any time period—a significant advantage over current reading-only glass thermometers. Range is -58 to 158°F and -50 to 70°C with a resolution of 1° and accuracy of ±1°C.

Alarm feature provides alert when temperature rises above or falls below a set point. Alarm is programmable in 1° increments. Visual and audible signals continue even if temperature returns to non-alarm range. Ten-foot micro-cable permits refrigerator doors to close on it.

Solid-state probe eliminates mercury contamination in refrigerators. Probe is sealed in a miniature bottle (1 x 2-1/2 inches) filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration). Eliminates concerns about contact with food or drinking water.

Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO/IEC 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Technology).
Customers Who Bought This Also Bought