Order Entry
Canada
ContactUsLinkComponent
VWR® Traceable® Extreme-Accuracy Digital Thermometers
Catalogue # 89094-758
Fournisseur:  VWR International
undefined
VWR® Traceable® Extreme-Accuracy Digital Thermometers
Catalogue # 89094-758
Fournisseur:  VWR International

Spécifications

  • Description:
    Extreme-Accuracy Digital Thermometer
  • Temperature resolution:
    0.01°
  • W×D×H:
    70×108×19 mm (23/4 × 41/4 × 3/4")
  • Weight:
    141.7 g (5 oz.)
  • Cat. no.:
    89094-758
  • Calibration Points:
    0 °C (32 °F), 25 °C (77 °F), 37 °C (98.6 °F)
  • Accuracy:
    ±0.3 °C, ±0.05 °C within ±2 °C of 0.00 °C, 25.00 °C, 37.00 °C
  • Range:
    –50…+70 °C

Spécifications

À propos de cet article

Absolute high-precision thermometers provide the exact readings necessary for critical requirements, quality control checks, and routine measurements.

  • Traceable
  • Temperature in °C and °F
  • Stainless steel probe
  • Continuous visual and audio alerts of temperature changes
  • One-button self-calibration

These thermometers are ideal for monitoring temperatures in water baths, cuvettes, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater

The stainless steel probe can be used in liquids, semi-solids, and air/gas. Provides accurate readings when probe is immersed only 8mm (5/16"). Probe diameter: 3mm (1/8"). Probe stem length: 15.9 cm (61/4"). Overall probe length: 22.9 cm (9"). Thermometers feature one-button self-calibration. Display case is made of high-impact, chemical-resistant ABS plastic.

Display simultaneously shows maximum, minimum, and current probe temperatures, as well as room temperature. Alarm feature provides continuous visual and audio alerts when temperature rises above or falls below a programmable set point, even if temperature returns to non-alarm range.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with two AA alkaline batteries, stand, wall mount, Velcro® mounting strips, magnetic mounting strips, and stainless steel probe with 1.5m (5') cable.
Frequently Bought Together