ContactUsLinkComponent
VWR JAR SS CLEAR PC 250ML CS12
VWR® TraceClean® Wide Mouth Jars, Glass
# de catalogue 89094-002
Fournisseur:  VWR International
Small Business Enterprise
VWR JAR SS CLEAR PC 250ML CS12
VWR® TraceClean® Wide Mouth Jars, Glass
# de catalogue 89094-002
Fournisseur:  VWR International
Small Business Enterprise
Spécifications
  • Cap Size:
    58-400
  • Cap Type:
    Solid-Top
  • Capacity:
    250 ml (8.5 oz.)
  • Description:
    Processed (PC)
Spécifications
A propos de cet article
Type III (soda-lime) jars are for collection of solid matrices or liquids dispensed from bailers or peristaltic tubing.
  • Soda-lime glass fabrication
  • Wide-mouthed design
  • Available with solid-top or open-top caps
  • Individually numbered for traceability
  • Jars are available with solid-top or open-top caps
Processed (PC) and Quality-Assured® (QA) jars follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA jars include a Certificate of Analysis in every case.
QA jars also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) jars do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Solid-top caps with fluoropolymer resin liner are for use in semi-volatiles, pesticides, and PCBs; and select inorganic compounds. Open-top caps with ultrasonically bonded fluoropolymer resin/silicone septa are for use in volatile organic analysis.
Customers Who Bought This Also Bought