Order Entry
ContactUsLinkComponent
STOPPER GRND JNT PYREX 24/40
PYREX® Hollow Full Length [ST] Stoppers, Corning
# de catalogue 89091-610
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
STOPPER GRND JNT PYREX 24/40
PYREX® Hollow Full Length [ST] Stoppers, Corning
# de catalogue 89091-610
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®

Spécifications

  • ST Joint No.:
    24/40
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. no.:
    89091-610
  • Height Above Ground Joint:
    40 mm (1⁹⁄₁₆")
  • Supplier No.:
    7570N-24

Spécifications

A propos de cet article

Hollow, full length stopper closed at the bottom from PYREX®, borosilicate glass.

  • Interchangeable with standard taper joints

They are lightweight, but strong enough to keep chipping and breakage to a minimum. Barrel-shaped head is easy to clean and rotate, side grooves prevent slippage, and flat top allows stopper to stand on head.

Customers Who Bought This Also Bought