PYREX® Reaction Vial, Corning
# de catalogue 89091-560
Fournisseur: Corning
Spécifications
- Cap included:Yes
- Capacity:1.00 ml
- Description:Reaction vial
- Material:PYREX® Glass
- Cat. no.:89091-560
- Bottom Style:Conical
- Supplier No.:9900-1
Spécifications
A propos de cet article
Conical Reaction Vials serve as a replacement part for the 6949M series of PYREX® Microchemistry Kits.
Customers Who Bought This Also Bought