Order Entry
ContactUsLinkComponent
PIPET VOL CLS A ACCURED 100 ML
PYREX® Volumetric Pipettes, Color-Coded, Class A, Corning
# de catalogue 89091-078
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
PIPET VOL CLS A ACCURED 100 ML
PYREX® Volumetric Pipettes, Color-Coded, Class A, Corning
# de catalogue 89091-078
Fournisseur:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®

Spécifications

  • Capacity:
    100 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. no.:
    89091-078
  • Tolerance:
    ±0.08 ml
  • Band Color:
    Yellow
  • Supplier No.:
    7100-100

Spécifications

A propos de cet article

Colored markings remain sharply legible for the life of pipet.

  • Reusable, rugged construction
  • Color-coded
  • Colored enamel graduations and markings

Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.

Customers Who Bought This Also Bought