ContactUsLinkComponent
Centrifuge Tube 500Ml Cs36
Corning® Centrifuge Tubes, Sterile, Polypropylene, Corning
Catalog # 89091-000
Supplier:  Corning
Product available on GSA Advantage®
Centrifuge Tube 500Ml Cs36
Corning® Centrifuge Tubes, Sterile, Polypropylene, Corning
Catalog # 89091-000
Supplier:  Corning
Product available on GSA Advantage®
Specifications
  • Volume:
    500 ml
  • Cat. no.:
    89091-000
  • Maximum RCF:
    6000×g
  • Supplier No.:
    431123
Specifications
About this item
PP, with HDPE plug seal cap, sterile. Polyetherimide cushions.
  • Moulded graduations, conical bottom
  • Tubes are sterile and certified non pyrogenic
  • Support cushions must be used unless the rotor has appropriately shaped V-bottom holders
Ideal for applications requiring large volume centrifugation.
They are perfect for large volume centrifugation applications. They incorporate plug seal caps and have molded graduations.
Ordering information: Non sterile cushion sold separately.
Packaging: Bulk, 6/bag.
Customers Who Bought This Also Bought