ContactUsLinkComponent
Insert Pipet 1-200U Cs960
Corning® Pipette Tips, Universal, Corning
Catalog # 89089-958
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Insert Pipet 1-200U Cs960
Corning® Pipette Tips, Universal, Corning
Catalog # 89089-958
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • Volume:
    1 - 200 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Tip Color:
    Yellow
  • Packed:
    10 inserts, 96 each
  • Cat. no.:
    89089-958
  • Sterile:
    -
  • Supplier No.:
    4710
Specifications
About this item
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
  • Beveled orifice helps ensure accurate fluid delivery
  • All tips are certified RNase-/DNase-free and non pyrogenic
  • Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
  • Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Customers Who Bought This Also Bought