Specifications
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:89089-924
- Membrane Material:Polycarbonate (PC)
- Membrane Pore Size:0.4 µm
- Supplier No.:3391
Specifications
About this item
Permeable supports for HTS (high throughput screening) applications are excellent for ADMET screening, drug transport, and other assays.
The HTS Transwell® 96-well permeable support features 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit. Available with polyester or polycarbonate membranes in a variety of pore sizes. The 0.143cm² well membrane area provides significantly more surface area than comparable systems. Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access. Plates are optimized for automation, with multichannel feeder ports, an improved gripping surface, and standard bar codes. The reservoir plate allows for the simultaneous feeding of 96 wells, and is supplied with a removable media stabilizer to minimize the risk of spills. The tissue culture-treated receiver plates isolate each well to enable 96 individual assays. Components are sterilized by gamma radiation.