Corning® Cell Culture Roller Bottles, Corning
# de catalogue 89089-838
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Cap Size:48 mm
- Cap Type:Easy Grip
- Culture area:850 cm²
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Oneonta
- Cat. no.:89089-838
- Packaging:22/Bag
- Neck O.D.:45 mm
- Supplier No.:431344
Spécifications
A propos de cet article
Virgin PS, sterile.
- One piece seamless construction
- All bottles have printed lot numbers to aid product traceability
- Sterilized by gamma radiation
- Certified non-pyrogenic
- Ribbed design provides twice the surface area with the same exterior dimensions
Roller bottles available with smooth or ribbed surface design for expanded culture area. All bottles are treated for optimal cell attachment. The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields. All bottles are graduated, unless otherwise stated.
Note: Corning recommends 0.2 to 0.3 ml of medium per cm² of growth area, and setting roller rack speeds to provide 0.5 to 1.0 rpm.
Customers Who Bought This Also Bought