Spécifications
- Format:F-bottom
- Color:Black
- Bottom Style:Solid
- Supplier No.:781900
Spécifications
A propos de cet article
Greiner Bio-One's non-binding microplate surfaces prevent unwanted non-specific binding.
- Nonsterile
- Free of RNase, DNase, and DNA-olymerase inhibitors
- Nonpyrogenic
- Include solid and film well bottoms
- Improved signal-to-noise ratio
This is especially advantageous for sensitive biochemical assays
All plates are RNase-, DNase-, and pyrogen-free, and also free of DNA-polymerase inhibitors.
Item numbers 89130-328 and 89089-586 come in HiBase plate design.
Low amounts of biomolecular binding can cause an undesirable increase in background, resulting in decreased signal-to-noise ratio. Characterized by low protein, DNA, RNA, and peptide binding properties, the non-binding microplate surfaces significantly increase assay sensitivity by reducing background and improving signal-to-noise ratio. Non-binding microplates are featured in 96 well and 384 well formats in black, white, and clear. Microplates include solid and µClear film well bottoms.