ContactUsLinkComponent
384 Well Plate
384-Well Non-Binding Microplate, Nonsterile, Greiner Bio-One
# de catalogue 89089-584
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
384 Well Plate
384-Well Non-Binding Microplate, Nonsterile, Greiner Bio-One
# de catalogue 89089-584
Fournisseur:  Greiner Bio-One
Product available on GSA Advantage®
Spécifications
  • Format:
    F-bottom
  • Color:
    Black
  • Bottom Style:
    Solid
  • Supplier No.:
    781900
Spécifications
A propos de cet article
Greiner Bio-One's non-binding microplate surfaces prevent unwanted non-specific binding.
  • Nonsterile
  • Free of RNase, DNase, and DNA-olymerase inhibitors
  • Nonpyrogenic
  • Include solid and film well bottoms
  • Improved signal-to-noise ratio
This is especially advantageous for sensitive biochemical assays
All plates are RNase-, DNase-, and pyrogen-free, and also free of DNA-polymerase inhibitors.
Item numbers 89130-328 and 89089-586 come in HiBase plate design.
Low amounts of biomolecular binding can cause an undesirable increase in background, resulting in decreased signal-to-noise ratio. Characterized by low protein, DNA, RNA, and peptide binding properties, the non-binding microplate surfaces significantly increase assay sensitivity by reducing background and improving signal-to-noise ratio. Non-binding microplates are featured in 96 well and 384 well formats in black, white, and clear. Microplates include solid and µClear film well bottoms.
Ordering information: Lid not included.
Customers Who Bought This Also Bought