Order Entry
ContactUsLinkComponent
NextGen V-Vials
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
# de catalogue 89085-418
Small Business Enterprise
NextGen V-Vials
NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton, DWK Life Sciences
# de catalogue 89085-418
Small Business Enterprise

Spécifications

  • Cap included:
    Yes
  • Cap Size:
    13-425
  • Cap Type:
    Solid-top
  • Capacity:
    1.00 ml
  • Color:
    Clear
  • Description:
    Vials with caps
  • Cat. no.:
    89085-418
  • Diameter ×H:
    13×44 mm
  • Supplier No.:
    W986254NG

Spécifications

A propos de cet article

Sturdy screw-thread vials have a conical interior bottom for maximum retrieval of content.

Vials can be used for small-scale reactions, centrifugation, storage, and shipping. They are ideal for use in the biochemical, pharmaceutical, and radiochemical industries. Fire-glazed bottom protects against glass particulates. Low-particulate packaging protects against contamination during transportation and storage. Clear vials are made from Wheaton 33* low extractable borosilicate glass to safeguard against a change in product pH caused by alkali, which can be extracted by ordinary glass. Amber vials are made from Wheaton 320 low extractable borosilicate glass. Caps are black, phenolic solid-top or open-top with fluoropolymer resin-faced 14B rubber liners.

Certifications: Clear vials conform to ASTM Type I, Class A, and USP Type I requirements. Amber vials conform to USP Type I requirements.
Ordering information: These vials do not have interior ground joints.
Customers Who Bought This Also Bought