24-Well No-Bottom Plates, Greiner Bio-One
# de catalogue 89085-338
Fournisseur: Greiner Bio-One
Product available on GSA Advantage®
Spécifications
- Color:Black
- Cat. no.:89085-338
- Supplier No.:662000-06
Spécifications
A propos de cet article
Plates are made from virgin polystyrene resin and molded without the bottom of the plate.
- Virgin polystyrene construction
- Designed without a bottom
- Chimney-well design prevents cross-contamination
- Durable
They feature chimney-well design to help prevent cross-contamination.
Customers Who Bought This Also Bought