Accessories for Reaction Vessel Heads, Ace Glass
# de catalogue 89084-140
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Ace-Thred Compression Fittings, Ace-Thred #18, NPT size 3/4"
- Cat. no.:89084-140
- Supplier No.:12871-18
Spécifications
A propos de cet article
These are Compression Fitting PTFE Inserts for PTFE reaction vessel heads.
Customers Who Bought This Also Bought