ContactUsLinkComponent
40ML POR D FILTER TUBE, 24/40 INNER JOINTS TOP & BOTTOM, TWO STOPCOCK INLETS
Schlenck Filter Tube with Double Sidearms and Inner Standard Taper Joints, Ace Glass Incorporated
# de catalogue 89083-442
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
40ML POR D FILTER TUBE, 24/40 INNER JOINTS TOP & BOTTOM, TWO STOPCOCK INLETS
Schlenck Filter Tube with Double Sidearms and Inner Standard Taper Joints, Ace Glass Incorporated
# de catalogue 89083-442
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Porosity:
    10–20 µm
  • Description:
    Schlenck Filter Tube with Double Sidearms and Inner Standard Taper Joints
  • Cat. no.:
    89083-442
  • ST Joint Size:
    24/40
  • Supplier No.:
    7774-14
  • Side Port:
    Two Glass Stopcocks
Spécifications
A propos de cet article
Schlenk filter tubes feature an integral glass filter disc at the tube's mid-line and two sidearms with either a glass stopcock or a 0-3 inlet valve
Paired top and bottom inner joints. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought