Schlenck Filter Tube with Double Sidearms and Inner Standard Taper Joints, Ace Glass Incorporated
# de catalogue 89083-442
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Porosity:10–20 µm
- Description:Schlenck Filter Tube with Double Sidearms and Inner Standard Taper Joints
- Cat. no.:89083-442
- ST Joint Size:24/40
- Supplier No.:7774-14
- Side Port:Two Glass Stopcocks
Spécifications
A propos de cet article
Schlenk filter tubes feature an integral glass filter disc at the tube's mid-line and two sidearms with either a glass stopcock or a 0-3 inlet valve
Paired top and bottom inner joints. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought