ContactUsLinkComponent
3/8" FLEXIBLE BEAM USED WITH 6472-157 CHUCK TO CONNECT 3/8" MOTOR SHAFT TO 28MMOD STIR ROD
Flexible Shaft Coupling, 28 mm, Ace Glass Incorporated
# de catalogue 89083-348
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
3/8" FLEXIBLE BEAM USED WITH 6472-157 CHUCK TO CONNECT 3/8" MOTOR SHAFT TO 28MMOD STIR ROD
Flexible Shaft Coupling, 28 mm, Ace Glass Incorporated
# de catalogue 89083-348
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Flexible Beam
  • Cat. no.:
    89083-348
  • Shaft Diameter:
    ⅜"
  • Supplier No.:
    6472-159
Spécifications
A propos de cet article
This is a coated steel flex-beam connector, for attaching directly to stir motor drive shaft.
Fits three different standard motor shaft sizes, top. Comes with 1/2" stainless steel pin, (bottom) that attaches to top of the 28 mm nylon chuck. Stir shaft diameter, 28 mm.
Customers Who Bought This Also Bought