Flexible Shaft Coupling, 28 mm, Ace Glass Incorporated
# de catalogue 89083-348
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Flexible Beam
- Cat. no.:89083-348
- Shaft Diameter:⅜"
- Supplier No.:6472-159
Spécifications
A propos de cet article
This is a coated steel flex-beam connector, for attaching directly to stir motor drive shaft.
Fits three different standard motor shaft sizes, top. Comes with 1/2" stainless steel pin, (bottom) that attaches to top of the 28 mm nylon chuck. Stir shaft diameter, 28 mm.
Customers Who Bought This Also Bought