ContactUsLinkComponent
20MM DRILLED CAP, 10.5MM HOLE, NO LINER, CS/48
Micro Reaction Vial Caps, Ace Glass Incorporated
# de catalogue 89083-176
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
20MM DRILLED CAP, 10.5MM HOLE, NO LINER, CS/48
Micro Reaction Vial Caps, Ace Glass Incorporated
# de catalogue 89083-176
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Fits:
    GPI thread finish 20-400
  • Supplier No.:
    9590-49
Spécifications
A propos de cet article
Threaded cap with or without drilled hole (solid) which includes PTFE faced silicone rubber septum and one o-ring for making “O-Ring-CAP-SEAL.” For use with Microscale Reaction Vials.
Customers Who Bought This Also Bought