Micro Reaction Vial Caps, Ace Glass Incorporated
# de catalogue 89083-176
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Fits:GPI thread finish 20-400
- Supplier No.:9590-49
Spécifications
A propos de cet article
Threaded cap with or without drilled hole (solid) which includes PTFE faced silicone rubber septum and one o-ring for making “O-Ring-CAP-SEAL.” For use with Microscale Reaction Vials.
Customers Who Bought This Also Bought