ContactUsLinkComponent
19/22 ADAPTER CONNECTION HOSE W/ POR A FRITTED DISC.
Hose Connection Adapter, [ST], with Frit, Ace Glass Incorporated
# de catalogue 89082-638
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
19/22 ADAPTER CONNECTION HOSE W/ POR A FRITTED DISC.
Hose Connection Adapter, [ST], with Frit, Ace Glass Incorporated
# de catalogue 89082-638
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Hose Connection [ST] Adapter with Frit
  • Cat. no.:
    89082-638
  • Hose Connections:
    5/16" (A)
  • Joint Size:
    [ST] 19/22
  • Supplier No.:
    9069-116
Spécifications
A propos de cet article
These adapters feature a straight hose connection at top and contain a fritted disc.
  • Bottom ground [ST] inner joint
  • Top straight hose connection
  • Fritted disc
  • Fabricated from borosilicate glass
Featuring a ground standard taper inner joint at the bottom and a straight hose connection at the top. Adapters contain an integral Porosity A (145–174 micron) fritted disc.
Customers Who Bought This Also Bought