Hose Connection Adapter, [ST], with Frit, Ace Glass Incorporated
# de catalogue 89082-638
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Hose Connection [ST] Adapter with Frit
- Cat. no.:89082-638
- Hose Connections:5/16" (A)
- Joint Size:[ST] 19/22
- Supplier No.:9069-116
Spécifications
A propos de cet article
These adapters feature a straight hose connection at top and contain a fritted disc.
- Bottom ground [ST] inner joint
- Top straight hose connection
- Fritted disc
- Fabricated from borosilicate glass
Featuring a ground standard taper inner joint at the bottom and a straight hose connection at the top. Adapters contain an integral Porosity A (145–174 micron) fritted disc.
Customers Who Bought This Also Bought