ContactUsLinkComponent
ADAPTER, 28/15 SOCKET JOINT, 10MM TUBE END
Adapter, Outer Socket Joint, Ace Glass Incorporated
# de catalogue 89082-554
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
ADAPTER, 28/15 SOCKET JOINT, 10MM TUBE END
Adapter, Outer Socket Joint, Ace Glass Incorporated
# de catalogue 89082-554
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Outer Socket Joint Adapter
  • Cat. no.:
    89082-554
  • SJ Joint No.:
    25/15
  • Tube Size:
    10 mm
  • Ace-thred no.:
    -
  • Supplier No.:
    5221-05
Spécifications
A propos de cet article
This series of adapters is designed for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
  • Outer socket joints
  • Various Ace-Thred and tube ends
  • Fabricated from borosilicate glass
Adapters feature 28/15 and 35/25 sockets with various Ace-Thred and tube ends.
Customers Who Bought This Also Bought