ContactUsLinkComponent
TIP FILTER EDEK 1000 CS768
OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
# de catalogue 89082-363
Fournisseur:  Sorenson Bioscience
TIP FILTER EDEK 1000 CS768
OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
# de catalogue 89082-363
Fournisseur:  Sorenson Bioscience
Spécifications
  • Volume:
    50 - 1250 µl
  • Packed:
    e•dek™ Environmental Paperboard Racks
  • Filter:
    +
  • Tip Style:
    Graduated (200 µL, 500 µL, 1000 µL)
  • Supplier No.:
    10460
Spécifications
A propos de cet article
These pipette tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft.
  • Dual-shot tip design
  • Easy-to-seal
  • Color-coded collar
The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Packaging: Available in traditional plastic racks, or edek™ paperboard racks that fit seamlessly into paper recycling programs.
Customers Who Bought This Also Bought