OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
# de catalogue 89082-363
Fournisseur: Sorenson Bioscience
Spécifications
- Volume:50 - 1250 µl
- Packed:e•dek™ Environmental Paperboard Racks
- Filter:+
- Tip Style:Graduated (200 µL, 500 µL, 1000 µL)
- Supplier No.:10460
Spécifications
A propos de cet article
These pipette tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft.
- Dual-shot tip design
- Easy-to-seal
- Color-coded collar
The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Packaging: Available in traditional plastic racks, or edek™ paperboard racks that fit seamlessly into paper recycling programs.
Customers Who Bought This Also Bought