Bath Oil, Ace Glass
# de catalogue 89078-210
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Numéro CAS:
Spécifications
- Color:Clear
- Description:Bath Oil, Low-Temp
- Volume:0.9 L (30³⁄₇ oz.)
- Supplier No.:14115-05
Spécifications
A propos de cet article
An extremely stable, medium viscosity silicone heating bath oil
Will not show discoloration after 40 hours at maximum temperature.
Ordering information: Available in two temperature ranges: Low Temp — maximum 180°C; or High Temp — maximum 230°C. Low temp oil color is CLEAR. High temp oil color is AMBER.
Customers Who Bought This Also Bought