Glass-Filled Polypropylene Tube Connectors, Ace Glass Incorporated
# de catalogue 89077-408
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Connector Tube to MNPT (with 9.5 mm Pipe Thread)
- Cat. no.:89077-408
- Fits Tubing O.D.:12.7 mm (½")
- Supplier No.:12707-13
Spécifications
A propos de cet article
These glass-filled polypropylene connectors are designed for use with stainless steel, nylon, polyolefin, aluminum, and all other tubing.
Customers Who Bought This Also Bought